MAN238928833 Manchester Organics

4-Chloro-3-(trifluoromethyl)benzyl chloride

  • MDL number MFCD03412205
  • Molecular Weight : 229.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/J19026
Availability details for
Quantity
  • Availability
  • Add to Cart