MAN238928834 Manchester Organics

5-Chloro-2-fluoro-4-methylpyridine

  • MDL number MFCD04972410
  • Molecular Weight : 145.562

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F19023
Availability details for
Quantity
  • Availability
  • Add to Cart