MAN238928845 Manchester Organics

4-Fluoro-3-methylbenzotrifluoride

  • MDL number MFCD07778352
  • Molecular Weight : 178.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B14001
Availability details for
Quantity
  • Availability
  • Add to Cart