MAN238928871 Manchester Organics

4-Ethyl-5-fluoropyrimidine

  • MDL number MFCD06657566
  • Molecular Weight : 126.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U19365

Safety Information

MSDS https://www.manchesterorganics.com/U19365
Availability details for
Quantity
  • Availability
  • Add to Cart