MAN238928874 Manchester Organics

Ethyl 3-bromopyridine-4-carboxylate

  • MDL number MFCD11226640
  • Cas Number : 13959-01-8
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-bromoisonicotinate
Inchi Code 1S/C8H8BrNO2/c1-2-12-8(11)6-3-4-10-5-7(6)9/h3-5H,2H2,1H3
Inchi Key KAJIRIIDTCRPKH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/X19061
Availability details for
Quantity
  • Availability
  • Add to Cart