MAN238928893 Manchester Organics

2-Cyano-6-(trifluoromethyl)pyridine

  • MDL number MFCD07368048
  • Cas Number : 887583-52-0
  • Molecular Weight : 172.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/J19072
Availability details for
Quantity
  • Availability
  • Add to Cart