MAN238928913 Manchester Organics

4-(Trifluoromethyl)phenyl isocyanate

  • MDL number MFCD00002028
  • Molecular Weight : 187.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A19112

Safety Information

MSDS https://www.manchesterorganics.com/A19112
Availability details for
Quantity
  • Availability
  • Add to Cart