MAN238928919 Manchester Organics

1,3-Dichloro-2-(3,3,3-trifluoropropyl)benzene

  • MDL number MFCD11226648
  • Cas Number : 1099597-55-3
  • Molecular Weight : 243.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-dichloro-2-(3,3,3-trifluoropropyl)benzene
Inchi Code 1S/C9H7Cl2F3/c10-7-2-1-3-8(11)6(7)4-5-9(12,13)14/h1-3H,4-5H2
Inchi Key XITDEMGBNBNGIE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/L14033
Availability details for
Quantity
  • Availability
  • Add to Cart