MAN238928927 Manchester Organics

1-Fluoro-2-(2,2,2-trifluoroethyl)-4-(trifluoromethyl)benzene

  • MDL number MFCD11226656
  • Cas Number : 1099597-68-8
  • Molecular Weight : 246.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-fluoro-2-(2,2,2-trifluoroethyl)-4-(trifluoromethyl)benzene
Inchi Code 1S/C9H5F7/c10-7-2-1-6(9(14,15)16)3-5(7)4-8(11,12)13/h1-3H,4H2
Inchi Key PACQMIHUZCLGNX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/V14041
Availability details for
Quantity
  • Availability
  • Add to Cart