MAN238928936 Manchester Organics

1-Chloro-2-fluoro-3,4-bis-(trifluoromethyl)benzene

  • MDL number MFCD11226663
  • Cas Number : 1099597-43-9
  • Molecular Weight : 266.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-chloro-2-fluoro-3,4-bis(trifluoromethyl)benzene
Inchi Code 1S/C8H2ClF7/c9-4-2-1-3(7(11,12)13)5(6(4)10)8(14,15)16/h1-2H
Inchi Key RSGXVUJZLLOLMX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/C14050
Availability details for
Quantity
  • Availability
  • Add to Cart