MAN238928945 Manchester Organics

6-Chloro-2-fluoro-3-methylbenzotrifluoride

  • MDL number MFCD11226667
  • Cas Number : 1099597-48-4
  • Molecular Weight : 212.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-chloro-3-fluoro-4-methyl-2-(trifluoromethyl)benzene
Inchi Code 1S/C8H5ClF4/c1-4-2-3-5(9)6(7(4)10)8(11,12)13/h2-3H,1H3
Inchi Key OXDBCVFGACYJMJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/G14054
Availability details for
Quantity
  • Availability
  • Add to Cart