MAN238928953 Manchester Organics

1-Bromo-3-(3,3,3-trifluoropropyl)benzene

  • MDL number MFCD11226672
  • Cas Number : 1099597-60-0
  • Molecular Weight : 253.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/P14061
Availability details for
Quantity
  • Availability
  • Add to Cart