MAN238928973 Manchester Organics

2-Amino-5-methoxypyrimidine

  • MDL number MFCD00187864
  • Cas Number : 13418-77-4
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methoxy-2-pyrimidinamine
Inchi Code 1S/C5H7N3O/c1-9-4-2-7-5(6)8-3-4/h2-3H,1H3,(H2,6,7,8)
Inchi Key KAHHAPNRIQLSFT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z19111
Availability details for
Quantity
  • Availability
  • Add to Cart