MAN238928981 Manchester Organics

2,4-Dichloro-5-cyanopyrimidine

  • MDL number MFCD09746248
  • Cas Number : 3177-24-0
  • Molecular Weight : 173.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/J19120
Availability details for
Quantity
  • Availability
  • Add to Cart