MAN238928998 Manchester Organics

1,2,3,4,5-Pentafluoro-6-(2,2,2-trifluoroethyl)benzene

  • MDL number MFCD11226681
  • Cas Number : 27059-24-1
  • Molecular Weight : 250.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,2,3,4,5-pentafluoro-6-(2,2,2-trifluoroethyl)benzene
Inchi Code 1S/C8H2F8/c9-3-2(1-8(14,15)16)4(10)6(12)7(13)5(3)11/h1H2
Inchi Key AZSBRCDLVFIMRD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/W14067
Availability details for
Quantity
  • Availability
  • Add to Cart