MAN238929000 Manchester Organics

2,3-Difluoro-1,4-bis-(trifluoromethyl)benzene

  • MDL number MFCD11226683
  • Cas Number : 1099597-67-7
  • Molecular Weight : 250.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/U14065
Availability details for
Quantity
  • Availability
  • Add to Cart