MAN238929020 Manchester Organics

2-Bromo-4-methylbenzotrifluoride

  • MDL number MFCD11226554
  • Cas Number : 121793-12-2
  • Molecular Weight : 239.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-methyl-1-(trifluoromethyl)benzene
Inchi Code 1S/C8H6BrF3/c1-5-2-3-6(7(9)4-5)8(10,11)12/h2-4H,1H3
Inchi Key ILHMYYRIQKCMIZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/E14004
Availability details for
Quantity
  • Availability
  • Add to Cart