MAN238929027 Manchester Organics

6-Chloro-1H-indazole

  • MDL number MFCD07774234
  • Cas Number : 698-25-9
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-1H-indazole
Inchi Code 1S/C7H5ClN2/c8-6-2-1-5-4-9-10-7(5)3-6/h1-4H,(H,9,10)
Inchi Key VUZQHUVRBPILAX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/L19145
Availability details for
Quantity
  • Availability
  • Add to Cart