MAN238929031 Manchester Organics

7-Fluoro-1H-indazole

  • MDL number MFCD09026976
  • Molecular Weight : 136.126

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R19149
Availability details for
Quantity
  • Availability
  • Add to Cart