MAN238929037 Manchester Organics

1-Fluoro-2,3-bis-(trifluoromethyl)benzene

  • MDL number MFCD11226555
  • Cas Number : 1099597-20-2
  • Molecular Weight : 232.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/F14005
Availability details for
Quantity
  • Availability
  • Add to Cart