MAN238929066 Manchester Organics

2,4-Bis(trifluoromethyl)acetophenone

  • MDL number MFCD00042495
  • Molecular Weight : 256.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V19508

Safety Information

MSDS https://www.manchesterorganics.com/V19508
Availability details for
Quantity
  • Availability
  • Add to Cart