MAN238929103 Manchester Organics

2-Amino-4-(trifluoromethyl)benzonitrile

  • MDL number MFCD04974124
  • Cas Number : 1483-54-1
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/X19203
Availability details for
Quantity
  • Availability
  • Add to Cart