MAN238929118 Manchester Organics

1-Chloro-4-(1,1-difluoroethyl)-2,5-difluorobenzene

  • MDL number MFCD11226702
  • Cas Number : 1138444-89-9
  • Molecular Weight : 212.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y14092
Availability details for
Quantity
  • Availability
  • Add to Cart