MAN238929122 Manchester Organics

3-(1H-Benzimidazol-2-yl)benzoic acid

  • MDL number MFCD02323224
  • Cas Number : 402944-81-4
  • Molecular Weight : 238.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N19218
Availability details for
Quantity
  • Availability
  • Add to Cart