MAN238929130 Manchester Organics

4-(1,1-Difluoroethyl)-1-fluoro-2-(trifluoromethyl)benzene

  • MDL number MFCD11226712
  • Cas Number : 1138445-20-1
  • Molecular Weight : 228.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(1,1-difluoroethyl)-1-fluoro-2-(trifluoromethyl)benzene
Inchi Code 1S/C9H6F6/c1-8(11,12)5-2-3-7(10)6(4-5)9(13,14)15/h2-4H,1H3
Inchi Key XJEPHGWSBZJRIX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/H14101
Availability details for
Quantity
  • Availability
  • Add to Cart