MAN238929134 Manchester Organics

2-(1,1-Difluoroethyl)-1-fluoro-4-(trifluoromethyl)benzene

  • MDL number MFCD11226715
  • Cas Number : 1022980-26-2
  • Molecular Weight : 228.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1,1-difluoroethyl)-1-fluoro-4-(trifluoromethyl)benzene
Inchi Code 1S/C9H6F6/c1-8(11,12)6-4-5(9(13,14)15)2-3-7(6)10/h2-4H,1H3
Inchi Key CRJZWEIYKUCTML-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/L14104
Availability details for
Quantity
  • Availability
  • Add to Cart