MAN238929157 Manchester Organics

2-Bromo-4-(1,1-difluoroethyl)-1-fluorobenzene

  • MDL number MFCD11845795
  • Cas Number : 1138444-85-5
  • Molecular Weight : 239.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/W14113
Availability details for
Quantity
  • Availability
  • Add to Cart