MAN238929175 Manchester Organics

2,5-Dichloro-3-methylpyridine

  • MDL number MFCD03840754
  • Cas Number : 59782-88-6
  • Molecular Weight : 162.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/A19231
Availability details for
Quantity
  • Availability
  • Add to Cart