MAN238929176 Manchester Organics

Tetrakis(4-nitrophenyl)methane

  • MDL number MFCD12546931
  • Cas Number : 60532-62-9
  • Molecular Weight : 500.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/C10972
Availability details for
Quantity
  • Availability
  • Add to Cart