MAN238929194 Manchester Organics

6-Chloropyridine-3,4-dicarboxylic acid

  • MDL number MFCD03411696
  • Cas Number : 243835-70-3
  • Molecular Weight : 201.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-3,4-pyridinedicarboxylic acid
Inchi Code 1S/C7H4ClNO4/c8-5-1-3(6(10)11)4(2-9-5)7(12)13/h1-2H,(H,10,11)(H,12,13)
Inchi Key LVQPMCFUDXELIS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/J15238
Availability details for
Quantity
  • Availability
  • Add to Cart