MAN238929201 Manchester Organics

1-(1,1-Difluoroethyl)-4-fluorobenzene

  • MDL number MFCD11845816
  • Molecular Weight : 160.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z14164
Availability details for
Quantity
  • Availability
  • Add to Cart