MAN238929235 Manchester Organics

1-Methyl-4-(pentafluoroethyl)benzene

  • MDL number MFCD09909908
  • Molecular Weight : 210.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G14148
Availability details for
Quantity
  • Availability
  • Add to Cart