MAN238929240 Manchester Organics

1-(Difluoromethyl)-2,4-difluorobenzene

  • MDL number MFCD06657968
  • Molecular Weight : 164.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R15434

Safety Information

MSDS https://www.manchesterorganics.com/R15434
Availability details for
Quantity
  • Availability
  • Add to Cart