MAN238929243 Manchester Organics

1-(Difluoromethyl)-2,4,5-trifluorobenzene

  • MDL number MFCD08276247
  • Molecular Weight : 182.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V15628

Safety Information

MSDS https://www.manchesterorganics.com/V15628
Availability details for
Quantity
  • Availability
  • Add to Cart