MAN238929252 Manchester Organics

6-Bromoimidazo[1,2-a]pyrazine

  • MDL number MFCD09750036
  • Cas Number : 912773-24-1
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D19257
Availability details for
Quantity
  • Availability
  • Add to Cart