MAN238929264 Manchester Organics

4-Chloro-3-methoxybenzoic acid

  • MDL number MFCD00269640
  • Molecular Weight : 186.59238

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S19269

Safety Information

MSDS https://www.manchesterorganics.com/S19269
Availability details for
Quantity
  • Availability
  • Add to Cart