MAN238929275 Manchester Organics

4-Methoxy-3,5-dimethyl-2-pyridinemethanol

  • MDL number MFCD01076195
  • Molecular Weight : 167.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X19700

Safety Information

MSDS https://www.manchesterorganics.com/X19700
Availability details for
Quantity
  • Availability
  • Add to Cart