MAN238929276 Manchester Organics

4-Cyano-5-methylsulfanylthiophene-2-carboxylic acid

  • MDL number MFCD12026743
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H10977
Availability details for
Quantity
  • Availability
  • Add to Cart