MAN238929294 Manchester Organics

4-Chloro-7-fluoroquinazoline

  • MDL number MFCD08236729
  • Molecular Weight : 182.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N19431

Safety Information

MSDS https://www.manchesterorganics.com/N19431
Availability details for
Quantity
  • Availability
  • Add to Cart