MAN238929309 Manchester Organics

1-Chloro-4-(1,1-difluoroethyl)-2-(trifluoromethyl)benzene

  • MDL number MFCD11845801
  • Cas Number : 1138444-94-6
  • Molecular Weight : 244.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-chloro-4-(1,1-difluoroethyl)-2-(trifluoromethyl)benzene
Inchi Code 1S/C9H6ClF5/c1-8(11,12)5-2-3-7(10)6(4-5)9(13,14)15/h2-4H,1H3
Inchi Key XXRFAXKQLBZEOB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/C14167
Availability details for
Quantity
  • Availability
  • Add to Cart