MAN238929311 Manchester Organics

3-Bromo-4-cyanopyridine

  • MDL number MFCD00234143
  • Cas Number : 13958-98-0
  • Molecular Weight : 183.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/A19302
Availability details for
Quantity
  • Availability
  • Add to Cart