MAN238929312 Manchester Organics

6-Nitro-5-methyl(1H)indazole

  • MDL number MFCD09026996
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X19723

Safety Information

MSDS https://www.manchesterorganics.com/X19723
Availability details for
Quantity
  • Availability
  • Add to Cart