MAN238929317 Manchester Organics

3-Iodopyridine-4-carboxylic acid methyl ester

  • MDL number MFCD03411715
  • Cas Number : 188677-49-8
  • Molecular Weight : 263.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-iodoisonicotinate
Inchi Code 1S/C7H6INO2/c1-11-7(10)5-2-3-9-4-6(5)8/h2-4H,1H3
Inchi Key XZUYSNLMTTZFDY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/B19303
Availability details for
Quantity
  • Availability
  • Add to Cart