MAN238929332 Manchester Organics

4-Chloro-3-(trifluoromethoxy)benzyl alcohol

  • MDL number MFCD06660248
  • Molecular Weight : 226.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P19480

Safety Information

MSDS https://www.manchesterorganics.com/P19480
Availability details for
Quantity
  • Availability
  • Add to Cart