MAN238929335 Manchester Organics

6-Bromo-2-chloro-1,8-naphthyridine

  • MDL number MFCD07781152
  • Molecular Weight : 243.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Country of Origin GB
Product Link https://www.manchesterorganics.com/M10981
Availability details for
Quantity
  • Availability
  • Add to Cart