MAN238929398 Manchester Organics

4-(Morpholin-4-ylmethyl)benzoic acid

  • MDL number MFCD01167722
  • Molecular Weight : 221.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B19374

Safety Information

MSDS https://www.manchesterorganics.com/B19374
Availability details for
Quantity
  • Availability
  • Add to Cart