MAN238929401 Manchester Organics

4,5-Bis(trifluoromethyl)imidazole

  • MDL number MFCD12546953
  • Cas Number : 651-34-3
  • Molecular Weight : 204.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,5-bis(trifluoromethyl)-1H-imidazole
Inchi Code 1S/C5H2F6N2/c6-4(7,8)2-3(5(9,10)11)13-1-12-2/h1H,(H,12,13)
Inchi Key OTWRLZYPCYXBFT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/M15289
Availability details for
Quantity
  • Availability
  • Add to Cart