MAN238929405 Manchester Organics

2-(3-Phenoxyphenyl)propanoic acid

  • MDL number MFCD00072027
  • Cas Number : 31879-05-7
  • Molecular Weight : 242.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/G19354
Availability details for
Quantity
  • Availability
  • Add to Cart