MAN238929431 Manchester Organics

4-Cyano-5-methylsulfanylthiophene-2-carboxylic acid ethyl ester

  • MDL number MFCD01859680
  • Cas Number : 116170-84-4
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/G10976
Availability details for
Quantity
  • Availability
  • Add to Cart