MAN238929456 Manchester Organics

3-Cyclopentylproponal

  • MDL number MFCD09745006
  • Molecular Weight : 126.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H19497

Safety Information

MSDS https://www.manchesterorganics.com/H19497
Availability details for
Quantity
  • Availability
  • Add to Cart